Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ Cartridge
Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
1 x 1 mL
1 x 5 mL
Unit Size:
Each
Description
- Strep-Tactin™XT Superflow™ cartridges contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins with HPLC/FPLC devices like Äkta™ systems (Cytiva).
- In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
- In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
- As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
Specifications
Specifications
Binding Capacity | 5.7 mg protein/ mL resin |
Capacity (Metric) | 1 x 1 mL |
Quantity | 1 Cartridge |
For Use With (Application) | Protein Purification |
Sufficient For | Chromatpgraphy workstations with 10 to 32 connections |
For Research Use Only.