Learn More
Cole-Parmer™ Straight Compression Union
Offers very good chemical resistance. Ideal when working with any rigid tubing, including tubing made from fluoropolymers, other rigid plastics and metals
276.00 NOK - 482.00 NOK
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Min. Operating Temperature (English) -40°F
Min. Operating Temperature (Metric) -40°C
Sterilization Method Autoclave, Ethylene Oxide or Gamma
View More Specs
Products 4
Product Code Brand Tubing Size Price Quantity & Availability  
Product Code Brand Tubing Size Price Quantity & Availability  
11825413
View Documents
Cole-Parmer™
31406-01
1/8 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11835413
View Documents
Cole-Parmer™
31406-02
1/4 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11845413
View Documents
Cole-Parmer™
31406-03
3/8 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11815413
View Documents
Cole-Parmer™
31406-00
1/16 in.
482.00 NOK
Each
Only null left
Estimated Shipment: 25-07-2025
Add to basket
Description
Description
  • Very good chemical resistance.
  • Rates to 220psi at 77°F (25°C).
  • Usable from 32 to 212°F (0 to +100°C).
  • Compression fittings work well with rigid tubing such as PTFE or polypropylene.
  • A compression fitting has a nut that compresses rigid tubing against fitting body for high-pressure applications.
  • Compression connector family is ideal when working with rigid tubing.
  • When selecting your fittings, consider your tubing type, material compatibility and temperature and pressure conditions.
Specifications
Specifications
Autoclavable
-40°F
Autoclave, Ethylene Oxide or Gamma
200°F
Stainless Steel
-40°C
220 psi
93°C