Learn More
BRAND™ Stoppers for Centrifuge Tube
Polyethylene caps prevent centrifuge tubes from spilling during processing.
Brand:  BRAND™ 115376
298.32 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10662052
  • 307.00 NOK / Pack of 100
Only null left
Estimated Shipment: 05-09-2025
Description
  • Polyethylene construction
  • For use with polypropylene centrifuge tubes
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene centrifuge tubes
160mL
Caps
100/PK
Polyethylene