missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13579-1-AP-20UL
This item is not returnable.
View return policy
Description
The yeast ′Sterile 20′ gene (STE20) functions upstream of the mitogen-activated protein kinase (MAPK) cascade. In mammals, protein kinases related to STE20 can be divided into 2 subfamilies based on their structure and regulation. Members of the PAK subfamily (see PAK3; MIM 300142) contain a C-terminal catalytic domain and an N-terminal regulatory domain that has a CDC42 (MIM 116952)-binding domain. In contrast, members of the GCK subfamily (see MAP4K2; MIM 603166), also called the Sps1 subfamily, have an N-terminal catalytic domain and a C-terminal regulatory domain without a CDC42-binding domain. STK24 belongs to the GCK subfamily of STE20-like kinases.Specifications
STK24 | |
Polyclonal | |
Unconjugated | |
STK24 | |
MST 3, MST3, MST3B, STE20, STE20 like kinase MST3, STK24, STK3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
223255, 361092, 8428 | |
-20°C | |
Liquid |
Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B0LT89, Q99KH8, Q9Y6E0 | |
Stk24 | |
STK24 Fusion Protein Ag4483 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction