Learn More
Lenz Laborglasinstrumente™ Stirrer Bearings with a Dynamic Double Seal
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Stirrer Bearings with a Dynamic Double Seal are ideal for use with applications under vacuum or under slight overpressure.
4135.00 NOK - 4305.00 NOK
Specifications
Diameter (Metric) | 16 mm |
---|---|
Material | Borosilicate Glass |
Material (Seal) | PTFE |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11913931
|
Lenz Laborglasinstrumente™
06306206 |
Joint (NS) 45/46 |
4305.00 NOK
Each |
Estimated Shipment: 02-09-2025
Log in to see stock. |
|||||
11993921
|
Lenz Laborglasinstrumente™
06306204 |
Joint (NS) 29/32 |
4135.00 NOK
Each |
Estimated Shipment: 02-09-2025
Log in to see stock. |
|||||
Description
- With extended guidance for reduced vibrations and a longer lifetime
- Made of borosilicate glass
- PTFE(sealings)
Specifications
16 mm | |
Borosilicate Glass | |
PTFE |