Learn More
Sampling Systems™ SteriWare™ Micro-Spatulas
Allows samples to be taken from very small containers such as vials with its narrow sampling blade. Sampling Systems™ SteriWare™ Micro-Spatulas are ideal for handling small quantities of powders and granules.
Brand: Sampling Systems™ 8015N-170S
1567.60 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Different blade widths available. Ideal for small vials or sample containers
- Also available in HDPE for extra chemical resistance
Specifications
Polystyrene | |
Single-use | |
White | |
6 mm |
0.5 mL | |
Sterile | |
170 mm |