Learn More
Sigma Laborzentrifugen™ Steel Centrifuge Tables
Use these centrifuge tables for a wide variety of applications. Sigma™ Steel Centrifuge Tables are made of sheet steel with two doors and space for accessories.
Brand: Sigma Laborzentrifugen™ 17927
1338.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- With 2 doors
- Manufactured from varnished sheet steel
- With space for accessories
- Movable on lockable castors
Specifications
Centrifuge Table | |
Sigma 3-16, Sigma 3-18 | |
515 mm |
Steel | |
490mm W x 684mm D x 515mm H | |
490 mm |