Learn More
Schneider Gerd™ Stand Ring with Socket
€ 20.05 - € 25.15
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
10746475
|
Schneider Gerd™
2811 |
70 x 190 x 8 mm | N/A | ||||||
10756475
|
Schneider Gerd™
2812 |
100 x 220 x 8 mm | N/A | ||||||
10736475
|
Schneider Gerd™
2810 |
50 x 170 x 8 mm | N/A | ||||||
10285154
|
Schneider Gerd™
2813 |
130 x 250 x 8 mm | N/A | ||||||
Specifications
Socket | |
Stand Ring | |
1 Ea. |