Learn More
Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths
Combine with any Grant Heating Circulator or Bath. Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths has a temperature range of –15 to 200°C.
Brand:  Grant Instruments™ ST38
 View more versions of this product
Product Code. 12737779
  • 17620.00 NOK / Each
Only null left
Estimated Shipment: 11-08-2025
Description
Combine with any Grant Heating Circulator or Bath. Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths has a temperature range of –15 to 200°C.
  • High efficiency stainless steel construction
  • Templerature range: -15 to 200°C
  • Includes integral raised feet for ease of carrying, repositioning, and retort access
TRUSTED_SUSTAINABILITY
Specifications
Specifications
38 L
15°C to 200°C
730 mm
Optima range immersion thermostats - T100, TC120, TXF150 or TXF200
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Tank and Bridge Plate for Baths > Capacity: 38L