Learn More
Grant Instruments™ Stainless Steel Sloping Lids
Recommended for use above 60°C. Stainless Steel Sloping Lid avoids contamination, reduces evaporation and saves energy.
2730.00 NOK - 3890.00 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11436427
|
Grant Instruments™
LU28 |
SAP18, ST12, ST26 |
3375.00 NOK
Each |
Estimated Shipment: 30-10-2025
Log in to see stock. |
|||||
11446427
|
Grant Instruments™
LU36 |
SAP34, ST38 |
3890.00 NOK
Each |
Estimated Shipment: 30-10-2025
Log in to see stock. |
|||||
11426427
|
Grant Instruments™
LU14 |
SAP12, JBN12, SBB12 |
3000.00 NOK
Each |
Estimated Shipment: 30-10-2025
Log in to see stock. |
|||||
11416427
|
Grant Instruments™
LU6 |
SAP5, ST5 |
2730.00 NOK
Each |
Estimated Shipment: 30-10-2025
Log in to see stock. |
|||||
Description
- For use with Sub Aqua Pro Series Unstirred Water Bath
- Triangular-shaped stainless steel with plastic handle
- Choose from the following available sizes:
- 5L (LU6)
- 12L (LU14)
- 18 or 26L (LU28)
- 34L (LU36)
Specifications
Stainless Steel |