Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN750plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SN750plusB02075
3345.85 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17558735
  • 3435.00 NOK / Each
Only null left
Estimated Shipment: 17-09-2025
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SN750plus Sterilizer
Drip Tray
Rim: 15 mm