Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SF75, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SF75B02072
982.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17578395
  • 1010.00 NOK / Each
Only null left
Estimated Shipment: 08-08-2025
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SF75 Sterilizer
Drip Tray
Rim: 15 mm