Learn More
Schneider Gerd™ Stainless Steel Flexible Micro-spatula
Use this spatula for a variety of applications. Schneider Gerd™ Stainless Steel Flexible Micro-spatulas are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 372
23.26 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10634575
  • 73.00 NOK / Each
Only null left
Estimated Shipment: 01-10-2025
Description
  • Stainless steel spatula
  • Ideal for working with powders, stirring and mixing
  • Perfect for miniscule amounts of chemicals
Specifications
Specifications
1 Ea.
Powder Patel Flexible Micro-Spatula
Stainless Steel
Powder Patel
Steel
150 mm