Learn More
Welch Ilmvac™ Stainless Steel KF Flexible Tube
Brand:  Welch Ilmvac™ 710761
1151.16 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10542353
  • 1185.00 NOK / Each
Only null left
Estimated Shipment: 15-09-2025
Specifications
Stainless Steel
1 Each
KF Flexible Tube
50 cm
DN 16
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Welch Ilmvac™ Stainless Steel KF Flexible Tube > Size: DN 16