Learn More
Agencinox™ Stainless Steel Funnels
870.00 NOK - 893.00 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11791923
|
Agencinox™
4220 |
1.5 L |
893.00 NOK
Each |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||||
11972068
|
Agencinox™
4215 |
0.5 L |
870.00 NOK
Each |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
Specifications
Stainless Steel | |
With handle |
Product Title
Agencinox™ Stainless Steel Funnels