Learn More
BINDER™ Stacking Adapter
Stacking Adapter for use with CB 160 Incubators.
Brand: BINDER™ 9051-0035
462.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with CB 160 Incubators
Specifications
Stacking adapter for the thermally isolated stacking of two BINDER CO2 incubators | |
BINDER CB 160 Incubator | |
Steel | |
Adapter |