Learn More
Nabertherm™ Square Saggar
Square saggar for furnaces LHTC and LH.
Brand:  Nabertherm™ 699000279
1754.82 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15330345
  • 1800.00 NOK / Each
Only null left
Estimated Shipment: 12-09-2025
Description
  • Tmax: 1600°C
The load is placed in ceramic saggars for optimal utilization of the furnace space. Up to three saggars can be stacked on top of each other in the furnace. Each saggar has cut-outs for better ventilation. The top saggar should be closed with a lid made of ceramics also.
Specifications
Specifications
1600°C
Square Saggar