Learn More
Julabo™ Spring Clamps for Shaking Water Bath
Brand:  Julabo™ 8970607
46.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10094270
  • 47.15 EUR / Each
Only null left
Estimated Shipment: 29-09-2025
Specifications
Holds 300 mL Erlenmeyer flask
SW22, SW23 shaking water bath
Spring Clamp