Learn More
Julabo™ Spring Clamps for Shaking Water Bath
Brand: Julabo™ 8970606
45.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Holds 200 to 250 mL Erlenmeyer flasks | |
SW22, SW23 shaking water bath | |
Spring Clamp |