Learn More
JP Selecta™ Spatula with Spoon and Flat Ends
Suitable for general laboratory purpose. JP Selecta™ Spatula with Spoon and Flat Ends is made of AISI 304 stainless steel.
Brand:  JP Selecta™ 1096120
 View more versions of this product
Product Code. 12037554
  • 10.00€ / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Description
  • Made of AISI 304 stainless steel
Specifications
Specifications
1 Pk.
18 mm
Stainless Steel (304)
Flat End/Spoon Ended Spatula
120 x 18 mm
120 mm