Learn More
SP110 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11502-1-AP-150UL
 View more versions of this product
Product Code. 16868623
  • 4796.50 NOK / 150µL
Description
Susceptibility to tuberculosis (TB) in mice has recently been attributed to the IPR1 gene. IPR1 is a member of the SP100/SP140 family of nuclear body proteins and encodes a leukocyte-specific nuclear body component. The protein can function as an activator of gene transcription and may serve as a nuclear hormone receptor coactivator. Alternative splicing has been observed for this gene and three transcript variants, encoding distinct isoforms, have been identified. SP110 is the closest homolog of the IPR1 protein in humans. The IPR1/Sp110 gene product might play a role in integrating signals generated by intracellular pathogens with mechanisms controlling innate immunity, cell death, and pathogenesis. IPR1/Sp110 is up-regulated after infection with M. tuberculosis and required by Anaplasma phagocytophilum for infection of human promyelocytic cells. Defects in Sp110 are a cause of severely impaired resistance to infection by M. tuberculosis.
Specifications
Specifications
SP110
Polyclonal
Unconjugated
SP110
IFI41, IFI75, IPR1, SP110, SP110 nuclear body protein, Speckled 110 kDa, VODI
Rabbit
Antigen Affinity Chromatography
RUO
109032, 3431
-20°C
Liquid
Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BVK9, Q9HB58
SP110
SP110 Fusion Protein Ag2042
150 μL
Primary
Human, Mouse