Learn More
Bel-Art™ SP Scienceware™ Micro-Mill™ Grinder Blades
Replacement blades for MicroMill grinder
366.00 NOK
Specifications
Product Type | Replacement Blade |
---|---|
For Use With (Application) | Ideal for processing minerals, pharmaceuticals, pigment blends and other solid samples |
For Use With (Equipment) | MicroMill grinder |
Speed | 12,000 rpm |
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
11316354
|
Bel-Art™
H37257-0000 |
Stainless Steel | N/A | ||||||
Description
Stainless-steel or hard-faced blades.
Certifications
CE Marked
Specifications
Replacement Blade | |
Ideal for processing minerals, pharmaceuticals, pigment blends and other solid samples | |
MicroMill grinder | |
12,000 rpm |