Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Brand: Thermo Scientific™ SK1830D
15091.43 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Maximize capacity on the Thermo Scientific™ Solaris Orbital Shakers with universal platforms and a variety of clamps to fit most vessels.
Certifications
UL, cUL, CE
Services
1 Year
Specifications
Dual Stack Universal Platform | |
18 x 30 in. | |
457.2 mm | |
610mm | |
Solaris 4000 | |
1 Year |
Certificate of Analysis (COA), Certificate of Conformance (COC) | |
18 in. | |
30 in. | |
Solaris 18x30 Dual Stack Universal platform | |
Two platforms, screws, and supporting rods |