Learn More
Memmert™ Software AtmoCONTROL for UN160plus
AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand:  Memmert™ UN160plusFDAQ1
29937.18 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17519935
  • 30690.00 NOK / Each
Only null left
Estimated Shipment: 13-08-2025
Description
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
UN160plus Oven
Software