Learn More
Sodium Sulfite, ≥98.0%, ACS Reagent, Honeywell Fluka™
Product Code. 15672450 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
ACS Reagent,≥98.0%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]
Specifications
Melting Point 600°C
Color White
Quantity 2.5 kg
Linear Formula Na2SO3
UN Number NONH for all modes of transport
Solubility Information Soluble in water, 126g/L at 20°C
Formula Weight 126.04g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder or Crystals
Density 2.6g/cm3
Chemical Name or Material Sodium Sulfite