Learn More
Sodium Sulfide Hydrate, Honeywell™
Product Code. 15681440 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
for gravimetric analysis
  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
PubChem CID 45051681
SMILES [Na+].[Na+].[S--]
Specifications
Color Colorless
Quantity 1 kg
UN Number UN1849
IUPAC Name disodium sulfanediide
Formula Weight 78.04g/mol (anhydrous basis)
Percent Purity 59 to 65%
Packaging Glass Bottle
Appearance Flakes
Chemical Name or Material Sodium Sulfide Hydrate