Learn More
Sodium Peroxide, puriss. p.a. ACS Reagent, Honeywell Fluka™
Product Code. 15603440 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 1 kg
Puriss. p.a., ACS Reagent, Reag. ISO,≥95%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]
Specifications
Quantity 6 x 1 kg
UN Number UN1504
Formula Weight 77.98
Percent Purity ≥95%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Peroxide, Reag. ISO