Learn More
Sodium hypophosphite monohydrate, 97+%, water ca 12-17%
Product Code. p-7232608
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
CAS: 10039-56-2 | H4NaO3P | 105.99 g/mol
Sodium hypophosphite monohydrate is used for electroless nickel plating agent and water treatment agent. It is also used in petroleum fields. Further, it is used as a food additive. In addition, it reduces nickel ions in solution to metallic nickel on metal substrates as well as on plastic substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS 10039-56-2
Molecular Formula H4NaO3P
Molecular Weight (g/mol) 105.99
MDL Number MFCD06252379
InChI Key PLZNPHDJGFDNRM-UHFFFAOYSA-M
Synonym Sodium phosphinite monohydrate;
IUPAC Name sodium hydrate oxophosphinite
SMILES O.[Na+].[O-][PH2]=O
Specifications
Physical Form Solid
Quantity 500 g
Percent Purity ≥97%