missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 97+%

Product Code. p-4407862
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases.Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in the ammonia borane dehydrogenation. Sodium borohydride is used as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces the aldehyde and keto groups to give the corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium tetrahydridoborate
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Melting Point 400°C (decomposition)
Color White
Physical Form Granules
Quantity 100 g
Assay Percent Range >97+%
Solubility Information Soluble in water,ammonia and amines. Insoluble in ether.
Formula Weight 37.83
Sensitivity Moisture sensitive
Appearance White
Density 1.074 g/mL
Chemical Name or Material Sodium borohydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium borohydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium borohydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.