Learn More
Sodium Bicarbonate, ≥99.5%, Reagent Grade, Honeywell™
Product Code. 15608130 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Poly bottle
Reagent Grade, powder,≥99.5%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O
Specifications
Color Colorless
pH 8.0 to 8.6 (at 20°C)
Quantity 1 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly bottle
Appearance Powder
Density 2.2g/cu.cm at 20°C
Chemical Name or Material Sodium bicarbonate