Learn More
Sodium 100/Potassium 100mmol/L, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Certificate of Analysis | NIST Certificate |
Concentration | 100m mol/L |