Learn More
Socorex™ Acura™ Single-channel Micropipette Volumetric Module
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ Single-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your single-channel micropipetting needs.
923.00 NOK - 1135.00 NOK
Specifications
Number of Channels | 1 |
---|---|
Product Line | Acura |
Type | Electronic Pipette Volumetric Module |
For Use With (Equipment) | Acura™ Electro 926 Micropipette |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11743276
|
Socorex™
800.0010 |
0.5 to 10 μL |
1135.00 NOK
Each |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||||
11773276
|
Socorex™
800.1000 |
50 to 1000 μL |
923.00 NOK
Each |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||||
11753276
|
Socorex™
800.0050 |
2.5 to 50 μL |
923.00 NOK
Each |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||||
11763276
|
Socorex™
800.0200 |
10 to 200 μL |
923.00 NOK
Each |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||||
Description
- One of 27 available volumetric assemblies.
- Increase versatility and save costs.
- Disassembles and reassembles without tools.
- Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
1 | |
Acura | |
Electronic Pipette Volumetric Module | |
Acura™ Electro 926 Micropipette |