Learn More
Saint Gobain Life Sciences Silicon Folding Skirt Stopper
Use this stopper for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Silicon Folding Skirt Stopper is particularly suited for all types of tissue culture work.
394.00 NOK - 749.00 NOK
Specifications
Color | Transparent |
---|---|
Material | Silicone |
Product Type | Folding Skirt Stopper |
Quantity | 1 Ea. |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11744598
|
Saint Gobain Life Sciences
467007-100 |
7.1 mm |
749.00 NOK
Pack of 100 |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
11714608
|
Saint Gobain Life Sciences
467018-20 |
16 mm |
394.00 NOK
Pack of 20 |
Estimated Shipment: 05-09-2025
Log in to see stock. |
|||||
Description
- Two-part folding skirt stoppers
- Designed with a double seal to keep container contents secure
- Ideal for securing glass, metal and plastic containers
- The lower section grips the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim
- Upper hollow plug section can be perforated to extract contents without removing the stopper and remains watertight even after several perforations
- Biologically neutral and non-toxic silicone caps for closing tubes and flasks
- Odorless, tasteless, non-adherent and resistant to aging
- Nontoxic, biologically neutral
- FDA/BGA approved
- For test tubes
- Easy to install
Specifications
Transparent | |
Silicone | |
Folding Skirt Stopper | |
1 Ea. |