Learn More
BINDER™ Left Access Port (10 to 30 mm)
For Use With: BINDER units ED 23, ED 400
Brand: BINDER™ 8012-0537
This item is not returnable.
View return policy
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 30 mm, left | |
Steel | |
Left | |
30 mm |
BINDER Chambers ED 23, ED 400 | |
Access Port | |
Silicone Plug |