Learn More
BRAND™ SILBERBRAND Compact Burette Spare Amber Burette Tube
Shop All Brand Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
Description
- Constructed of Borosilicate Glass 3.3
- Amber glass
Specifications
Specifications
Color | Amber |
Capacity (Metric) | 50 mL |
Length (Metric) | 69.5 cm |
For Use With (Equipment) | Compact Burette |
Material | Borosilicate Glass (5.4) |
Height (Metric) | 1.8 cm |