Learn More
Epredia™ Shandon™ Eosin-Y Stain
Product Code. p-4530257 Shop All Epredia Products
Click to view available options
Quantity:
1L
4L
Each
Type:
Alcoholic Eosin Y Stain
Aqueous Eosin Y Stain
Unit Size:
4 L
Pack of 2
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
  • Ready-to-use
  • Produces consistent results
  • Stable and precipitate-free; no filtering
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic or aqueous formulations
TRUSTED_SUSTAINABILITY
Specifications
Type Alcoholic Eosin Y Stain
Quantity 1L
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H350-May cause cancer.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
Supplemental information
  • MIXTURE LIST-Contains : ethyl, alcohol, methyl alcohol, isopropyl alcohol, acetic acid, eosin-Y dye