Learn More
Epredia™ Cytoclip™ Stainless-Steel Slide Clip
The Epredia™ Cytoclip Stainless-Steel Slide Clip holds Cytofunnel™ disposable sample chamber against the microscope slide.
Brand:  Epredia™ 59910052
3637.81 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12608036
  • 11345.00 NOK / Pack of 6
Only null left
Estimated Shipment: 08-09-2025
Description
  • For preparation in the Epredia Shandon Cytospin™ Cytocentrifuge
  • Autoclavable and reusable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
6/Box
Side Clip