Learn More
SGMS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19050-1-AP-20UL
This item is not returnable.
View return policy
Description
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.Specifications
SGMS1 | |
Polyclonal | |
Unconjugated | |
SGMS1 | |
hmob33, MOB, MOB1, Protein Mob, SGMS1, SMS1, sphingomyelin synthase 1, TMEM23, Transmembrane protein 23 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
208449, 259230, 353229 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TSX5, Q86VZ5, Q8VCQ6 | |
Sgms1 | |
SGMS1 Fusion Protein Ag5246 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |