Learn More
SGEF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12493-1-AP-20UL
 View more versions of this product
Product Code. 16870234
  • 1696.32 NOK / 20µL
Description
Activates RhoG GTPase by promoting the exchange of GDP by GTP. Required for the formation of membrane ruffles during macropinocytosis. Required for the formation of cup-like structures during trans-endothelial migration of leukocytes. In case of Salmonella enterica infection, activated by SopB, which induces cytoskeleton rearrangements and promotes bacterial entry.
Specifications
Specifications
SGEF
Polyclonal
Unconjugated
ARHGEF26
CSGEF, HMFN1864, SGEF
Rabbit
Antigen Affinity Chromatography
RUO
26084
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96DR7
ARHGEF26
SGEF Fusion Protein Ag3198