missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGEF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12493-1-AP-150UL
This item is not returnable.
View return policy
Description
Activates RhoG GTPase by promoting the exchange of GDP by GTP. Required for the formation of membrane ruffles during macropinocytosis. Required for the formation of cup-like structures during trans-endothelial migration of leukocytes. In case of Salmonella enterica infection, activated by SopB, which induces cytoskeleton rearrangements and promotes bacterial entry.Specifications
SGEF | |
Polyclonal | |
Unconjugated | |
ARHGEF26 | |
CSGEF, HMFN1864, SGEF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26084 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96DR7 | |
ARHGEF26 | |
SGEF Fusion Protein Ag3198 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction