Learn More
Trajan™ Replacement Needle for 10μL Syringe
A range of quality needles to fit a SGE syringe in a range of lengths and tip styles
585.00 NOK - 1040.00 NOK
Specifications
Product Type | Replacement Needle |
---|---|
For Use With (Equipment) | Syringes (10μL) |
Product Code | Brand | Needle Point Style | Diameter (Metric) Inner | Diameter (Metric) Outer | Length (Metric) Needle | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Point Style | Diameter (Metric) Inner | Diameter (Metric) Outer | Length (Metric) Needle | Price | Quantity & Availability | ||
15228593
|
Trajan™
037011 |
Cone | 0.11 mm | 0.63 mm | 50 mm |
585.00 NOK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10166970
|
Trajan™
037610 |
On-column | 0.1 mm | 0.17 mm | 100 mm |
1040.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10176970
|
Trajan™
037675 |
On-column | 0.1 mm | 0.23 mm | 75 mm |
1040.00 NOK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Replacement Needle | |
Syringes (10μL) |