Learn More
SGE™ LC Pump Priming Syringe
For loop fill injection or sample loop flush
Brand: SGE™ 008762
1028.32 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Fixed Luer lock
Specifications
Used for loop fill injection or sample loop flush | |
5MDF-LL-GT | |
5 mL |
Manual Syringe | |
Ea. | |
LC |