Learn More
Cytiva Sentino™ Funnel Adapter
Accessory adapter for Sentino filter funnels.
Brand:  Cytiva 4891
Product Code. 17124441
  • 6670.00 NOK / Pack of 3
Only null left
Estimated Shipment: 13-04-2026
Description
  • For microbiology manifold
  • Lab manifold designed for flexibility and reduction in cross-contamination with interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel (316L)
Non-sterile
3/Pk.
Funnel Adapter