Learn More
DWK Life Sciences Screw Cap with Aperture, for safety standard ground sockets
For fixing the DURAN™ core in the DURAN™ threaded sleeve.
1.84CHF - 117.00CHF
Specifications
Quantity 10/Pk.
View More Specs
Products 6
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15951551
View Documents
DWK Life Sciences
286215704
NS 45/40
117.00CHF
Pack of 10
Only null left
Estimated Shipment: 23-07-2025
Add to basket
15921551
View Documents
DWK Life Sciences
286215207
NS 19/26
23.90CHF
Pack of 10
Only null left
In Stock
Add to basket
15911551
View Documents
DWK Life Sciences
286215001
NS 14/23
18.35CHF
Pack of 10
Only null left
Estimated Shipment: 30-07-2025
Add to basket
10040151
View Documents
DWK Life Sciences
286215301
NS 24/29 Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
N/A
15941551
View Documents
DWK Life Sciences
286215404
NS 29/32
40.90CHF
Pack of 10
Only null left
In Stock
Add to basket
15844608
View Documents
DWK Life Sciences
286215507
NS 34/35
61.30CHF
Pack of 10
Only null left
In Stock
Add to basket
Description
Description
The open screw cap reliably fixes the core of the DURAN™ security cuts in the sleeve. When the screw connection is opened, the ground joints are automatically separated from each other.
  • Reliable fixation of the DURAN™ safety sections in one another (core and sleeve)
  • When the screw connection is opened, the ground joints are automatically separated from each other
Specifications
Specifications
10/Pk.