Learn More
Kimberly-Clark Professional™ Scott™ Essential™ Examination Couch Paper
Economic and portable, lightweight roll. Scott™ Essential™ Examination Couch Paper ideal for protecting couches in the healthcare industry.
1470.00 NOK - 1835.00 NOK
Specifications
Product Type | Couch Cover |
---|---|
Color | White |
Material | Paper |
Disposable | Single-use |
Format | Wrapped Roll |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12600543
|
Kimberly-Clark Professional™
7160 |
38 x 39 cm |
1470.00 NOK
1 roll |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
12555518
|
Kimberly-Clark Professional™
7161 |
37 x 50 cm |
1835.00 NOK
6 rolls |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
Description
- Strong, tear-resistant and durable
- Compatible with dispenser 7056
Specifications
Couch Cover | |
Paper | |
Wrapped Roll |
White | |
Single-use |