Learn More
BRAND™ Schiefferdecker Staining Trough
Soda-lime glass construction, for 10 slides
Brand:  BRAND™ 472500
3258.10 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 11839580
  • 4135.00 NOK / Pack of 10
Estimated Shipment: 11-06-2025
Explore more special offers
Description
  • For 10 slides, size 76 x 26 mm
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Staining Trough
    10
    Documents
    One moment while we fetch your results.
    Product Title
    BRAND™ Schiefferdecker Staining Trough > Dimensions (L x W): 85 x 70 mm