Learn More
Buerkle™ Stainless Steel Sample Spoon
Buerkle™ Stainless Steel (AISI 304) Sample Spoon is a specially shaped spoon that allows you to reach almost every corner in a container and prevents samples from trickling out.
89.35 NOK - 255.00 NOK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10297412
|
Buerkle™
5320-0210 |
210 mm |
103.00 NOK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
10494612
|
Buerkle™
5320-0300 |
300 mm |
170.00 NOK
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
|||||
10552733
|
Buerkle™
5320-0150 |
150 mm |
89.35 NOK
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
|||||
10562733
|
Buerkle™
5320-0180 |
180 mm |
95.75 NOK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
10044162
|
Buerkle™
5320-0400 |
400 mm |
219.00 NOK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
16369541
|
Buerkle™
5320-0500 |
500 mm |
255.00 NOK
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
|||||
Description
- Made of AISI 304 (1.4301) stainless steel
- Largely chemical resistant (not against acids and alkalis)
Specifications
Stainless Steel (V2A) | |
Autoclavable |