Learn More
Saline-Sodium Phosphate-EDTA (SSPE), 20X Solution, Fisher BioReagents
455.00 NOK
Specifications
Concentration 2.9 to 3.1M
Form Liquid
Storage Requirements RT
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10499113
View Documents
Fisher BioReagents
BP1328-1
1L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Saline–Sodium Phosphate–EDTA (SSPE) is used in bacterial screening and hybridization procedures.
Specifications
Specifications
2.9 to 3.1M
Liquid
RT
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Saline-sodium phospate-EDTA 20X solution