Learn More
SCAT™ Safety Waste Caps, Electrostatic Conductive
SCAT Safety Waste Caps, made of PTFE-EL, are hermetically sealing caps for the safe disposal of your liquid HPLC waste. The SCAT exhaust filter with three layers of activated carbon binds vapors from solvents, acids and alkalis. It ensures safe pressure equalization and occupational safety.
Brand: SCAT™ 460060
1544.61 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Made of electrically conductive PTFE-EL material to prevent static discharge
- Available with sereral threat sizes and numbers capillaries ports (5 to 11.5 mm ID)
- GL14 connection for use with SCAT exhaust filters
Specifications
Black | |
PTFE-EC | |
Waste Cap |
2 x PFA fitting 2.3 mm OD, 2 x PFA fitting 3.2 mm OD, 1 x PP tube connector 5 to 11.5 mm ID, 1 x GL14 connection for exhaust filter | |
S60/61 |