Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Extension Funnel
Safety Waste Cap 'LISA', V3.0 Extension Funnel is an accessory for LISA HPLC/UHPLC systems.
925.00 NOK - 995.00 NOK
Specifications
For Use With (Equipment) | HPLC / UHPLC Systems |
---|---|
Material | PTFE |
Thread Size | M30x35 |
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
17346429
|
SCAT
350120 |
Not Electrically Conductive | Extension Funnel |
925.00 NOK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
||||
17376429
|
SCAT
450120 |
Electrically Conductive | Extension Funnel with Earthing Cable |
995.00 NOK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
||||
Description
- Accessory for Safety Waste Cap 'LISA,' V3.0
- Extension Funnel with Hinged Lid
- Made of PTFE for optimal chemical resistance
Specifications
HPLC / UHPLC Systems | |
PTFE | |
M30x35 |